missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Cassette II Slotted Tissue Cassettes in Tube Packs
Slotted Cassettes are ideal for routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.
128.00 € - 154.00 €
Specifications
Closure Included | Yes |
---|---|
Quantity | 750/Cs. |
Pore Style | Slots |
Closure Type | Separate Lid, Front Hinge |
PrintMate Compatible | Yes |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
16314056
|
Epredia™
B851729BE |
Beige |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16324056
|
Epredia™
B851729BL |
Blue |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16334056
|
Epredia™
B851729GN |
Green |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16344056
|
Epredia™
B851729GY |
Gray |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16354056
|
Epredia™
B851729LA |
Lavender |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16364056
|
Epredia™
B851729LG |
Light Green |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16374056
|
Epredia™
B851729LI |
Lilac |
154.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16384056
|
Epredia™
B851729OR |
Orange |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16394056
|
Epredia™
B851729PK |
Pink |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16304066
|
Epredia™
B851729TN |
Tan |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16314066
|
Epredia™
B851729WH |
White |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16324066
|
Epredia™
B851729YW |
Yellow |
128.00 €
Pack of 750 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
Slots | |
Yes | |
Cassette II Biopsy Cassette |
750/Cs. | |
Separate Lid, Front Hinge | |
For Use With PrintMate™ Printers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title