Learn More
Fisherbrand™ Catalyst Norprene Food Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
285.00 € - 2315.00 €
Product Code | Brand | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Thickness (Metric) | Price | Quantity & Availability | ||||
15511335
|
Fisherbrand™
|
0.8 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15521335
|
Fisherbrand™
|
1.6 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15531335
|
Fisherbrand™
|
3.2 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15541335
|
Fisherbrand™
|
4.8 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15551335
|
Fisherbrand™
|
6.4 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15561335
|
Fisherbrand™
|
8 mm | 1.6 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15571335
|
Fisherbrand™
|
4.8 mm | 2.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15581335
|
Fisherbrand™
|
6.4 mm | 2.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15591335
|
Fisherbrand™
|
8 mm | 2.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15501345
|
Fisherbrand™
|
9.5 mm | 2.4 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Available in Platinum-Cured Silicone, TPEs, and PVC
- Ensures optimal amount of tubing is in pump head
Specifications
Stopped tubing assembly |