Learn More
Fisherbrand™ Catalyst PharMed BPT Two-Stop Precision Tubing Sets
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™ 97100-27
 View more versions of this product
Product Code. 15571365
  • 138.00 € / Pack of 12
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Preconfigured ready-to-use links and assemblies.
  • Time-saving assembly
  • For large cartridges
  • Four formulations for range of chemical compatibility
  • Use with Catalyst extension coils/tubing
Specifications
Specifications
Two-stop precision tubing set
4.8 mm