Learn More
Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Stopped Tubing Assembly
Preconfigured ready-to-use links and assemblies.
Brand:  Fisherbrand™ 97001-73
 View more versions of this product
Product Code. 15531355
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Preconfigured ready-to-use links and assemblies.
  • Available in Platinum-Cured Silicone, TPEs, and PVC
  • Ensures optimal amount of tubing is in pump head
Specifications
Specifications
Stopped Tubing Assembly
4.8 mm
1.6 mm