Learn More
Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Stopped Tubing Assembly
Description
Preconfigured ready-to-use links and assemblies.
- Available in Platinum-Cured Silicone, TPEs, and PVC
- Ensures optimal amount of tubing is in pump head
Specifications
Stopped Tubing Assembly | |
6.4 mm | |
6.4mm |