missing translation for 'onlineSavingsMsg'
Learn More

Catechol-O,O-diacetic acid, 97%, Thermo Scientific Chemicals

35.70 € - 124.00 €

Chemical Identifiers

CAS 5411-14-3
Molecular Formula C10H10O6
Molecular Weight (g/mol) 226.184
MDL Number MFCD00014353
InChI Key PPZYHOQWRAUWAY-UHFFFAOYSA-N
Synonym 1,2-phenylenedioxydiacetic acid, o-phenylenedioxydiacetic acid, 2,2'-1,2-phenylenebis oxy diacetic acid, catechol-o,o-diacetic acid, 2-2-carboxymethoxy phenoxy acetic acid, 2,2'-1,2-phenylenebis oxy bisacetic acid, 2-carboxymethoxy phenoxyacetic acid, 2,2'-benzene-1,2-diylbis oxy diacetic acid, acmc-1alcz, catechol-o,o'-diacetic acid
PubChem CID 79427
IUPAC Name 2-[2-(carboxymethoxy)phenoxy]acetic acid
SMILES C1=CC=C(C(=C1)OCC(=O)O)OCC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381417
View Documents
Thermo Scientific Alfa Aesar
L01843.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11391417
View Documents
Thermo Scientific Alfa Aesar
L01843.18
50 g
124.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5411-14-3
226.184
PPZYHOQWRAUWAY-UHFFFAOYSA-N
79427
C1=CC=C(C(=C1)OCC(=O)O)OCC(=O)O
C10H10O6
MFCD00014353
1,2-phenylenedioxydiacetic acid, o-phenylenedioxydiacetic acid, 2,2'-1,2-phenylenebis oxy diacetic acid, catechol-o,o-diacetic acid, 2-2-carboxymethoxy phenoxy acetic acid, 2,2'-1,2-phenylenebis oxy bisacetic acid, 2-carboxymethoxy phenoxyacetic acid, 2,2'-benzene-1,2-diylbis oxy diacetic acid, acmc-1alcz, catechol-o,o'-diacetic acid
2-[2-(carboxymethoxy)phenoxy]acetic acid

Specifications

5411-14-3
C10H10O6
2057228
PPZYHOQWRAUWAY-UHFFFAOYSA-N
2-[2-(carboxymethoxy)phenoxy]acetic acid
79427
97%
177°C to 181°C
MFCD00014353
1,2-phenylenedioxydiacetic acid, o-phenylenedioxydiacetic acid, 2,2'-1,2-phenylenebis oxy diacetic acid, catechol-o,o-diacetic acid, 2-2-carboxymethoxy phenoxy acetic acid, 2,2'-1,2-phenylenebis oxy bisacetic acid, 2-carboxymethoxy phenoxyacetic acid, 2,2'-benzene-1,2-diylbis oxy diacetic acid, acmc-1alcz, catechol-o,o'-diacetic acid
C1=CC=C(C(=C1)OCC(=O)O)OCC(=O)O
226.184
226.18
Catechol-O,O-diacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 226-488-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only