missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Catechol Violet

Product Code. 11465517
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11465517

Brand: Thermo Scientific Chemicals B23513.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-41-3 | C19H14O7S | 386.37 g/mol

Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.

Solubility
Soluble in water, ethylene glycol, monomethyl ether and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-41-3
Molecular Formula C19H14O7S
Molecular Weight (g/mol) 386.37
MDL Number MFCD00005868
InChI Key RRRCKIRSVQAAAS-UHFFFAOYSA-N
PubChem CID 66993
IUPAC Name 4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
SMILES OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1

Specifications

Beilstein 353938
Solubility Information Soluble in water,ethylene glycol,monomethyl ether and ethanol.
Formula Weight 386.38
Quantity 25 g
Chemical Name or Material Catechol Violet

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.