Learn More
Applied Biosystems™ Cathode Buffer Container for 3500 Dx/3500xL Dx Genetic Analyzers
The CBC is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label.
Brand:  Applied Biosystems™ 4408258
Product Code. 15968826
  • 270.00 € / Pack of 4
Edge
Explore more special offers
Description
Description
The cathode buffer container (CBC) contains 1x running buffer to support all electrophoresis applications on the 3500 Dx series Genetic Analyzers.
Order Info
4 Pack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DNA, RNA
3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
4 each
Each package contains 4 buffer containers.
Store at 2 - 8°C.
Cathode Buffer Container
Wet Ice