Learn More
Applied Biosystems™ Cathode Buffer Container (CBC), for 3500/SeqStudio™ Flex
GREENER_CHOICE
The cathode buffer container (CBC) contains 1X running buffer to support all electrophoresis applications on Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.
Brand:  Applied Biosystems™ 4408256
Product Code. 12070120
  • 261.00 € / Pack of 4
Explore more special offers
Description
Description
The cathode buffer container (CBC) contains 1X running buffer to support all electrophoresis applications on Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers. The container has two separate compartments, the left side provides the cathode buffer for electrophoresis and the right side provides for a capillary wash and spent polymer waste ejection functionality between injections.
The CBC is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the CBC is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.
Order Info
4 Pack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Contains 4 packs of cathode buffer container. Store the Cathode Buffer Container at 2-8°C until ready to use. Refer to the expiration date on the label.
Cathode Buffer Container
4 pack
3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Sustainable packaging
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2411047 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number4408256
Lot Number2411046 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number4408256
Lot Number2410045 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number4408256
Lot Number2410044 Certificate TypeCertificate of Analysis Date09/01/2025 Catalog Number4408256
Lot Number2410043 Certificate TypeCertificate of Analysis Date27/12/2024 Catalog Number4408256
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ Cathode Buffer Container (CBC), for 3500/SeqStudio™ Flex >