missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CCL4 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Marque: Abnova™ H00006351-T01.100uL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
293T | |
CCL4 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 10.2kDa | |
Transient overexpression cell lysate was tested with anti-CCL4 antibody (Mfr. No. H00006351-D01P) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
NM_002984.2 | |
pCMV-CCL4 full-length | |
100 μL | |
Human |