missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CCND1-FGF19/CEN11p FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
1515.00 € - 2495.00 €
Specifications
Content And Storage | Store at 4°C in the dark. |
---|---|
Description | DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Format | Tube |
For Use With (Application) | FISH-Ce |
Gene ID (Entrez) | 595/9965 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013295
|
Abnova™
FG0185.100uL |
100 μL |
1515.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16023295
|
Abnova™
FG0185.200uL |
200 μL |
2495.00 €
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark. | |
Tube | |
595/9965 | |
Liquid |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ CCND1-FGF19/CEN11p FISH Probe