Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J305, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Brand:  Paragon Scientific CL11
Product Code. 16080172
  • 93.30 € / 500mL
Explore more special offers
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Manufactured in the United Kingdom
  • 2 year shelf life
Specifications
Specifications
Glass Amber Bottle
Yes
Liquid
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • CCS Viscosity Standard, CL110 (CL11)
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : dec-1-ene
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.