Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J324, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Brand:  Paragon Scientific CL48
Product Code. 16000030
  • 101.00 € / 500mL
Explore more special offers
Description
Description
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Full Traceability to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Manufactured in the United Kingdom
  • 2 year shelf life
Specifications
Specifications
Glass Amber Bottle
Yes
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.