Learn More
Gibco™ CD 293 Medium (1X)

Descripción
CD 293 Medium is a chemically-defined, protein-free medium optimized for the growth of suspension cultures of 293 cells. CD 293 Medium contains no proteins or peptides of animal, plant, or synthetic origin. There are also no undefined lysates or hydrolysates in the formulation.
Formulated without L-glutamine for increased stability; add 20 mL of 200 mM L-glutamine or 20 mL of GlutaMAX™-I Supplement per 1,000 mL of medium prior to use.
A 293 Medium Master file has been submitted to the FDA. If you wish to cross-reference a Master file for CD 293 Medium, please contact Technical Services or your local Sales Representative.
Especificaciones
Especificaciones
| Cell Line | 293F, 293 (HEK) |
| Classification | Animal Origin-free, Low Protein, Serum-free |
| Concentration | 1 X |
| Form | Liquid |
| Product Type | 293 Medium |
| Serum Level | Serum-free |
| Without Additives | No Glutamine |
| Product Line | Gibco |
| Quantity | 1000 mL |
| Shipping Condition | Room Temperature |
| Mostrar más |
For Research Use Only. Not for use in diagnostic procedures.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.