missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ CD OptiCHO™ AGT™ Medium
CD OptiCHO™ AGT™ Medium
Brand: Gibco™ A1122205
988.00 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
CD OptiCHO™ medium is specifically designed to offer high performance and yield with recombinant CHO cells in a chemically defined fed-batch environment. The protein-free, animal origin free formulation is designed to provide consistency and reduces the need to screen for adventitious agents. CD OptiCHO™ medium builds on Gibco™ CD CHO’s legacy of consistent cell growth and titers, while offering up to 40% better titer and cell growth over select competitors. Thanks to its low osmolality and advanced CHO CD EfficientFeed™ Liquid Nutrient Supplement Kit feeding options, CD OptiCHO™ medium is capable of gaining 2-5 fold increases in fed-batch productivity over batch culture.When to Use CD OptiCHO™ Medium
When to Consider Another Gibco™ Medium
Consider CD FortiCHO™ medium when:
Consider CD CHO medium when:
Consider CD DG44 medium when:
Liquid Media Formulations
Gibco™ AGT™ media are also available in a liquid format that offers the same performance and formulation as the AGT™ media format. Gibco™ liquid media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their AGT™ media counterparts.
Design Your Own Media Specific to Your Needs
Whether it’s a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs. With Gibco™ cGMP Media Custom Services you can put the manufacturing power and years of technical experience of our R&D and Bioproduction Application Specialists to work for you.
Specifications
1 x 10 L, Storage 2-8°C Dark and Dry | |
CHO | |
Suspension Cell Culture | |
CD OptiCHO AGT Medium | |
Non-sterile | |
No Glutamine, No Phenol Red | |
Securitainer Capped Vial | |
Magnesium, Sodium Phosphate | |
Certified | |
10 L | |
Wet Ice |
CO2 | |
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Powder (AGT) | |
Serum-free | |
High Glucose, Sodium Pyruvate, Sodium Bicarbonate | |
Low | |
No antibiotics | |
AGT™, Gibco™ | |
10 L | |
Medium does not support wild type DG44 cells |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction