Learn More
Gibco™ CD OptiCHO™ Medium
Brand: Gibco™ 12681011
162.45 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
CD OptiCHO™ medium is specifically designed to offer high performance and yield with recombinant CHO cells in a chemically defined fed-batch environment. The protein-free, animal origin free formulation is designed to provide consistency and reduces the need to screen for adventitious agents. CD OptiCHO™ medium builds on Gibco™ CD CHO's legacy of consistent cell growth and titers, while offering up to 40% better titer and cell growth over select competitors. Thanks to its low osmolality and advancedCHO CD EfficientFeed™ Liquid Nutrient Supplement Kit feeding options, CD OptiCHO™ medium is capable of gaining 2-5 fold increases in fed-batch productivity over batch culture.
When to Use CD OptiCHO™ Medium
- You are using a transfected CHO cell line other than CHO K1, GS CHO or CHO-S™ cell lines
- Optimization of feeding strategies will be part of your base medium selection testing in an effort to maximize productivity
- Your CHO cell line is 'finicky,' i.e. hard to grow or adapt
When to Consider Another Gibco™ Medium
Consider CD FortiCHO™ medium when:
- The cell line you are using is a transfected CHO K1, GS CHO or CHO-S™ cell line
- Maximum batch culture cell densities and protein titers are needed
- You have time to adapt cells into a new medium in an effort to maximize titers
- Cell health at harvest is a priority for downstream processing
Consider CD CHO medium when:
- Your GS CHO cells are recommended to be grown in CD CHO medium
- Your cells have been successfully cultured in CD CHO medium in the past and your process is nearing a regulatory filing (DMF available)
Consider CD DG44 medium when:
- You use parental DG44 or DXB11 CHO cells (dhfr-)
Dry Media Formulations
Gibco™ liquid media are available in an AGT™ (Advanced Granulation Technology) dry media format that is designed to provide increased consistency and productivity across all stages of production from development to commercial manufacturing. AGT™ media are manufactured animal origin-free with the same complete, serum-free, protein-free, chemically-defined formulation and performance as their liquid media counterparts.
Design Your Own Media Specific to Your Needs
Whether it's a bench-level experiment or large scale biomanufacturing process, we offer specialized formats, packaging, and quality levels specific to your needs.
Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Animal Origin-free, Chemically-defined, Protein-free, Serum-free | |
Suspension Cell Culture | |
CD OptiCHO Medium | |
Sodium Pyruvate, Sodium Bicarbonate | |
Low | |
No antibiotics | |
1 L | |
Hamster |
CHO | |
1 X | |
Liquid | |
Serum-free | |
No Glutamine, No Phenol Red | |
Bottle | |
Gibco™ | |
Room Temperature |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.