Learn More
CD107b Mouse anti-Human, PE, Clone: H4B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech PE-65053-100TESTS
This item is not returnable.
View return policy
Description
The CD107b antibody from Proteintech is a mouse monoclonal antibody generated with human adherent peripheral blood cells of human CD107b. This antibody recognizes human antigen. The CD107b antibody has been validated for the following applications: IF, FC analysis. This antibody is pe.
Specifications
CD107b | |
Monoclonal | |
10 μL/Test | |
Mouse | |
Primary | |
Human |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
H4B4 | |
PE | |
100 Tests | |
3920 | |
IgG1 |