Learn More
CD117 (c-Kit) Rat anti-Mouse, PE, Clone: ACK2, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech PE-65094-100UG
This item is not returnable.
View return policy
Description
The CD117 (c-Kit) antibody from Proteintech is a rat monoclonal antibody generated with murine il-3 dependent mast cells of mouse CD117 (c-Kit). This antibody recognizes mouse antigen. The CD117 (c-Kit) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
CD117 (c-Kit) | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
ACK2 | |
PE | |
100 μg | |
16590 | |
IgG2b |