Learn More
CD11b, PE, clone: VIM12, Invitrogen™
Mouse Monoclonal Antibody
349.00 € - 1156.00 €
Specifications
Antigen | CD11b |
---|---|
Clone | VIM12 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10425853
|
Invitrogen
CD11B04 |
0.5 mL |
349.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10088292
|
Invitrogen
CD11B044 |
2 mL |
1156.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD11b is implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement coated particles. It is identical to CR3, the receptor for the iC3b fragment of the third complement component. It probably recognizes the RGD peptide in C3b. CD11b is also a receptor for fibrinogen, factor X, and ICAM1. It recognizes P1 and P2 peptides of fibrinogen gamma chain. The Mac1 CD11b antigen is present on macrophages, granulocytes, natural killer cells, and blood monocytes. CD11b is expressed on 8% of spleen cells, 44% of bone marrow cells, and less than 1% of thymocytes, and is commonly used as a microglial marker in nervous tissue.
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.Specifications
CD11b | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
P11215 | |
3684 | |
Human CD11b (Mac-1) | |
Primary | |
Store at 2 - 8 °C. | |
ITGAM |
VIM12 | |
Monoclonal | |
Liquid | |
ASR, ASR/IVD | |
PBS with BSA and 0.1% sodium azide | |
CD11B, CR3A, MAC-1, MAC1A, MGC117044, MO1A, SLEB6, Integrin alpha M | |
ITGAM | |
IgG1 | |
Purified | |
Antibody |