Learn More
CD11b Mouse anti-Human, Unconjugated, Clone: ICRF44, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65116-1-IG-100UG
Product Code. 17214636
  • 84.00 € / 100µg
Explore more special offers
Description
Description
Integrins are cell adhesion receptors that are heterodimers composed of non-covalently associated α and β subunits (PMID: 9779984). CD11b, also known as Integrin alpha M or CR3A, belongs to the integrin alpha chain family. CD11b forms an α/β heterodimer with CD18 (integrin β2). CD11b/CD18 is implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement-coated particles and pathogens (PMID: 9558116; 20008295). CD11b/CD18 is a receptor for the complement protein fragment iC3b, and is also a receptor for fibrinogen, factor X and ICAM1 (PMID: 2971974; 15485828).
Specifications
Specifications
CD11b
Monoclonal
0.5 mg/mL
Mouse
Primary
Human, Mouse, Rat
IgG1
Flow Cytometry
ICRF44
Unconjugated
100 μg
3684