missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD134 Rat anti-Mouse, CoraLite 488, Clone: OX-86, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL488-65136-100TESTS
This item is not returnable.
View return policy
Description
CD134, also known as OX40 and TNFRSF4, is a member of the TNFR-superfamily of receptors (PMID: 2828930; 9766631). It is a type I transmembrane protein predominantly expressed on activated T cells which include CD4 and CD8 T cells, Th2, Th1, and Th17 cells, as well as regulatory T cells (Tregs) (PMID: 20307208). CD134 is activated by its cognate ligand CD134L (OX40L) and functions as a T cell co-stimulatory molecule (PMID: 26215166). CD134-CD134L interactions have been proposed as a potential therapeutic target for treating autoimmune diseases, cancer and infectious disease (PMID: 26215166; 19426222).
Specifications
CD134 | |
Monoclonal | |
5 μL/Test | |
Rat | |
Primary | |
Human, Mouse, Rat | |
IgG1 κ |
Flow Cytometry | |
OX-86 | |
CoraLite 488 | |
100 Tests | |
22163 | |
Liquid |