missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD14 Isolation Kit for FABian™
IBA Lifesciences™ CD14 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
Brand: IBA Lifesciences 6-6017-001
This item is not returnable.
View return policy
Description
- The kit is used for the selection of CD14+ cells from buffy coat or whole blood in conjunction with the FABian™ system.
- It includes CD14 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
- Each kit is sufficient for one FABian™ run to isolate cells from 4.25mL buffy coat.
- Alternatively 9mL whole blood may be used.
Specifications
Cell Isolation Kit | |
Human Whole Blood | |
1 Kit, Human |
2°C to 8°C | |
Isolation of label-free cells |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction