Learn More
CD161f Rat anti-Mouse, BV605, Clone: C8, BD OptiBuild™
Rat Monoclonal Antibody
Brand:  BD Biosciences 743513
Product Code. 15838309
  • 745.00 € / Each
Explore more special offers
Description
Description
CD161f Monoclonal antibody specifically detects CD161f in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD161f
Monoclonal
0.2 mg/mL
Rat
RUO
Mouse
Purified
Flow Cytometry
C8
Brilliant Violet 605
50 μg
Primary
Refrigerate (2 to 8°C)
IgG1 κ