missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD161f Rat anti-Mouse, BV650, Clone: C8, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 743514
388.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD161f Monoclonal antibody specifically detects CD161f in Mouse samples. It is validated for Flow Cytometry.
Specifications
CD161f | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
C8 | |
Brilliant Violet 650 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction