missing translation for 'onlineSavingsMsg'
Learn More

CD161f Rat anti-Mouse, BV786, Clone: C8, BD OptiBuild™

Rat Monoclonal Antibody

Brand:  BD Biosciences 743516

388.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15868309

  • 389.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD161f Monoclonal antibody specifically detects CD161f in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD161f
Monoclonal
0.2 mg/mL
Rat
RUO
Mouse
Purified
Flow Cytometry
C8
Brilliant Violet 786
50 μg
Primary
Refrigerate (2 to 8°C)
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.