Learn More
CD18 Rat anti-Mouse, BB700, Clone: C71/16, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 742102
388.12 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The C71/16 monoclonal antibody specifically binds to mouse CD18 which is also known as the common β2 chain of LFA-1 (CD11a/CD18, αLβ2 integrin), Mac-1 (CD11b/CD18, αMβ2 integrin), and gp150, 95 (CD11c/CD18, αxβ2 integrin). CD18 is a type I transmembrane glycoprotein. Expression of CD18 is limited to leukocytes, where it is widely distributed in consort with the three integrin α chains (CD11a, CD11b, and CD11c). Among splenocytes, NK cells have the highest density of CD18, and T lymphocytes express a higher density than the remaining cells. The β2 integrins are important mediators of leukocyte-endothelium interactions.
The antibody was conjugated to BD Horizon™ BB700, which is part of the BD Horizon Brilliant™ Blue family of dyes. It is a polymer-based tandem dye developed exclusively by BD Biosciences. With an excitation max of 485 nm and an emission max of 693 nm, BD Horizon BB700 can be excited by the 488 nm laser and detected in a standard PerCP-Cy™5.5 set (eg, 695/40nm filter). This dye provides a much brighter alternative to PerCP-Cy5.5 with less cross laser excitation off the 405 nm and 355 nm lasers.BD Horizon Brilliant™ Blue 700 (BB700) is a dye that was exclusively developed by BD Biosciences as brighter alternative to PerCP-Cy5.5. This dye also has less cross laser excitation off the 405 nm laser, resulting in less spillover into the violet channels compared to PerCP-Cy5.5. Due to similar excitation and emission properties, BD Horizon BB700 and PerCP-Cy5.5 cannot be used simultaneously.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CD18 | |
C71/16 | |
Brilliant Blue 700 (BB700) | |
Itgb2, Integrin beta-2, ITB2, LAD, LCAMB, LFA-1/Mac-1/p150,95 subunit beta | |
Cell membrane glycoproteins from mouse T-cell lymphoma BW5147 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.