missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD1a Ab-2, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD1a Ab-2, Mouse Monoclonal Antibody.

148.92 € - 740.00 €

Specifications

Antigen CD1a Ab-2
Clone MTB1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12682237
View Documents
Epredia™
MS-746-S0
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12692237
View Documents
Epredia™
MS-746-S1
Concentrated 500 μL
610.00 €
500µL
Please to purchase this item. Need a web account? Register with us today!
12672237
View Documents
Epredia™
MS-746-S
Concentrated 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12662237
View Documents
Epredia™
MS-746-R7
Prediluted 7 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

At least five CD1 genes (CD1a, b, c, d, and e) have been identified. CD1 is expressed on cortical thymocytes, Langerhans cells, and dendritic cells. It is absent on mature peripheral blood T cells but intracytoplasmic expression is detected on activated T lymphocytes. CD1 proteins have been demonstrated to restrict T-cell response to non-peptide lipid and glycolipid antigens and play a role in non-classical antigen presentation.

Host Species: Mouse

Clone: MTB1

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: External domain

Immunogen: Full external domain of CD1a

Molecular Weight: 49 kDa

Positive Control: Thymus

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD1a Ab-2
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
IgG2a κ
MTB1
Monoclonal
Mouse
Hematopoietic
Full external domain of CD1a
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.