missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24 Mouse anti-Human, FITC, Clone: ML5, BD
Mouse Monoclonal Antibody
157.00 € - 385.00 €
Specifications
Antigen | CD24 |
---|---|
Clone | ML5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15830669
|
BD Biosciences
560992 |
25 Tests |
157.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15838158
|
BD Biosciences
555427 |
100 Tests |
385.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ML5 monoclonal antibody specifically binds to CD24. CD24 is a 35-45 kDa two-chain glycoprotein expressed on the surface of B cells, granulocytes and most B-cell lines. CD24 may play a role in the regulation of B-cell proliferation and maturation.
Specifications
CD24 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
ML5 | |
Monoclonal | |
Mouse | |
Human | |
IgG2a κ | |
Affinity Purified | |
Heat Stable Antigen Homologue (HSA); Ba-1; CD24A |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title