Learn More
CD24 Rat anti-Mouse, APC, Clone: M1/69, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech APC-65121-100UG
This item is not returnable.
View return policy
Description
The CD24 antibody from Proteintech is a rat monoclonal antibody generated with n/a of mouse CD24. This antibody recognizes mouse antigen. The CD24 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
CD24 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
Primary | |
Mouse |
Flow Cytometry | |
M1/69 | |
APC | |
100 μg | |
12484 | |
IgG2b |