Learn More
IBA Lifesciences CD28 Isolation Kit for FABian™
IBA Lifesciences™ CD28 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
Brand:  IBA Lifesciences 6-6014-010
 View more versions of this product
Product Code. 15899774
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • The kit is used for the selection of CD28+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
  • It includes CD28 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from up to 9mL whole blood or up to 6.25mL buffy coat.
Specifications
Specifications
Non-magnetic Cell Isolation Kit
2°C to 8°C
Isolation of label-free cells
10 Kits, Human