Learn More
CD3 Mouse anti-Human, APC, Clone: OKT3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech APC-65133-100TESTS
Product Code. 16880506
  • 43.00 € / 100 tests
Explore more special offers
Description
Description
The CD3 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD3. This antibody recognizes human antigen. The CD3 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
Specifications
CD3
Monoclonal
5 μL (0.125 μg)/Test
Mouse
Primary
Human
Flow Cytometry
OKT3
APC
100 Tests
916
IgG2a