missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human, CoraLite 488, Clone: SK7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-65148-100TESTS
This item is not returnable.
View return policy
Description
CD3 is a multimeric protein associated with the T-cell receptor (TCR) to form a complex involved in antigen recognition and signal transduction (PMID: 15885124). CD3 is composed of CD3γ, δ, ε, and ζ chains (PMID: 1826255). It is expressed by thymocytes in a developmentally regulated manner, T cells, and some NK cells (PMID: 3289580). The TCR recognizes antigens bound to major histocompatibility complex (MHC) molecules. TCR-mediated peptide-MHC recognition is transmitted to the CD3 complex, leading to the intracellular signal transduction (PMID: 11985657).
Specifications
CD3 | |
Monoclonal | |
5 μL/Test | |
Mouse | |
Primary | |
Human, Mouse, Rat | |
IgG1 κ |
Flow Cytometry | |
SK7 | |
CoraLite 488 | |
100 Tests | |
916 | |
Liquid |