missing translation for 'onlineSavingsMsg'
Learn More

CD30 (Reed-Sternberg Cell Marker) Ab-4 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD30 (Reed-Sternberg Cell Marker) Ab-4, Mouse Monoclonal Antibody.

124.44 € - 519.00 €

Specifications

Antigen CD30 Ab-4
Clone HRS4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12610327
View Documents
Epredia
MS-1857-P0
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12620327
View Documents
Epredia
MS-1857-P1ABX
Concentrated 100 μL
447.00 €
100µL
Please to purchase this item. Need a web account? Register with us today!
12630327
View Documents
Epredia
MS-1857-P1
Concentrated 500 μL
399.00 €
500µL
Please to purchase this item. Need a web account? Register with us today!
12600327
View Documents
Epredia
MS-1857-P
Concentrated 1 mL
519.00 €
1mL
Please to purchase this item. Need a web account? Register with us today!
12690327
View Documents
Epredia
MS-1857-R7
Prediluted 7 mL
267.00 €
7mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD30, a single chain glycoprotein, is synthesized as a 90 kDa precursor which is processed in the Golgi complex into a membrane-bound phosphorylated mature 105/120kDa glycoprotein. The CD30/Ki-1 antigen is expressed by mononuclear Hodgkin and multinucleated Reed-Sternberg cells in Hodgkin's disease, by the tumor cells of a majority of anaplastic large cell lymphomas, and by a varying proportion of activated T and B cells. About one third of the Ki-1 positive lymphomas lack the leukocyte common antigen (CD45).

Host Species: Mouse

Clone: HRS4

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Immunogen: Recombinant extracellular domain of CD30 protein

Molecular Weight: 120 kDa (mature), 95 kDa (precursor)

Positive Control: Hodgkin's lymphoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CD30 Ab-4
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgG1
HRS4
Monoclonal
Mouse
Oncogenesis
Recombinant extracellular domain of CD30 protein
Primary
Videos
SDS
Documents

Documents

Product Certifications