missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD31 Isolation Kit for FABian™

For automated positive cell separation. The Isolation Kit for FABian™ provides label-free target cells without the use of high affinity antibodies and magnetic beads

90.41 € - 718.00 €

Specifications

Product Type Non-magnetic Cell Isolation Kit
Content And Storage 2°C to 8°C
Donor Source Human Whole Blood or Buffy Coat
For Use With (Application) Isolation of label-free cells
Sufficient For 9 mL Whole Blood
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16245350
View Documents
IBA Lifesciences
6-6016-001
1 Kit, Human This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16255350
View Documents
IBA Lifesciences
6-6016-010
10 Kits, Human This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • The kit is used for the selection of CD31+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
  • It includes CD31 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from 9 ml whole blood.
  • Specifications

    Specifications

    Non-magnetic Cell Isolation Kit
    Human Whole Blood or Buffy Coat
    9 mL Whole Blood
    2°C to 8°C
    Isolation of label-free cells
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.