missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD31 Monoclonal Antibody (LCI-9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA182381
This item is not returnable.
View return policy
Description
Mouse anti Pig CD31 antibody, clone LCI-9 recognizes porcine Platelet endothelial cell adhesion molecule, also known as CD31 or PECAM-1.
CD31 (platelet endothelial cell adhesion molecule-1, PECAM-1) is an inhibitory coreceptor involved in regulation of T cell and B cell signaling by a dual immunoreceptor tyrosine-based inhibitory motif (ITIM) that upon associated kinases-mediated phosphorylation provide docking sites for protein-tyrosine phosphatases. CD31 is expressed ubiquitously within the vascular compartment and is located mainly at junctions between adjacent cells. N-terminal Ig-like domain of CD31 is responsible for its homophilic binding, which plays an important role in cell-cell interactions. CD31 is a multifunctional molecule with diverse roles in modulation of integrin-mediated cell adhesion, transendothelial migration, angiogenesis, apoptosis, negative regulation of immunoreceptor signaling, autoimmunity, macrophage phagocytosis, IgE-mediated anaphylaxis and thrombosis. It is one of key regulatory molecules in vascular system.
Specifications
CD31 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
Q95242 | |
Pecam1 | |
Porcine CD31/human IgGFc fusion protein. | |
RUO | |
396941 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
LCI-9 | |
Unconjugated | |
Pecam1 | |
C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule | |
Mouse | |
2 mL | |
Primary | |
Pig | |
Antibody | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CD31 Monoclonal Antibody (LCI-9), Invitrogen™ > 2 mL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction