missing translation for 'onlineSavingsMsg'
Learn More

CD34 (Clone QBEnd/10) Ab-1 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry procedures with Epredia™ CD34 (Endothelial Cell Marker) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-363-P0

 View more versions of this product

Product Code. 15230675

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD34, a single chain transmembrane glycoprotein, is selectively expressed on human lymphoid and myeloid hematopoietic progenitor cells. Staining for CD34 has been used to measure angiogenesis, which reportedly predicts tumor recurrence. Antibody to CD34 also reacts with vascular endothelial cells in normal tissues and in benign and malignant proliferations. This antibody is of value in the study of benign and malignant vascular tumors as well as characterization of leukemias.

Host Species: Mouse

Clone: QBEnd/10

Isotype: IgG1

Species Reactivity: Human. Does not react with dog and rat. Others not known

Immunogen: Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta

Molecular Weight: 105-120 kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD34 (Endothelial Cell Marker) Ab-1
Monoclonal
200μg/mL
Mouse
100 μL
Oncogenesis
Human
IgG1
Immunohistochemistry (Paraffin)
QBEnd/10
Unconjugated
Detergent solubilized vesicular suspension prepared from a perfusate of human term placenta
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.