Learn More
CD4 Mouse anti-Human, CoraLite 488, Clone: OKT4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-65134-100TESTS
Product Code. 16810676
  • 144.00 € / 100 tests
Explore more special offers
Description
Description
The CD4 antibody from Proteintech is a mouse monoclonal antibody generated with human peripheral t cells of human CD4. This antibody recognizes human antigen. The CD4 antibody has been validated for the following applications: FC analysis. This antibody is coralite™488.
Specifications
Specifications
CD4
Monoclonal
5 μL/Test
Mouse
Primary
Human
Flow Cytometry
OKT4
CoraLite 488
100 Tests
920
IgG2b