Learn More
CD4 Mouse, PerCP, Clone: SK3, enQuireBio™
Mouse Monoclonal Antibody
Brand:  EnquireBio QAB10-PCP-25Tests
 View more versions of this product
Product Code. 15941478
  • 275.00 € / 25 tests
Explore more special offers
Specifications
CD4
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
CD4
Protein A/G Purified
RUO
920
IgG1 κ
Flow Cytometry
SK3
PerCP
NP_001181945
Mouse
25 Tests
Primary
Store at 2-4°C.