Learn More
CD44 Mouse anti-Human, PE, Clone: F10-44-2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech PE-65063-100TESTS
Product Code. 16831676
  • 195.00 € / 100 tests
Explore more special offers
Description
Description
The CD44 antibody from Proteintech is a mouse monoclonal antibody generated with purified t cells from human lymph nodes of human CD44. This antibody recognizes human antigen. The CD44 antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
Specifications
CD44
Monoclonal
10 μL/Test
Mouse
Primary
Human
Flow Cytometry
F10-44-2
PE
100 Tests
960
IgG2a