Learn More
CD45.2 Mouse anti-Mouse, APC, Clone: 104, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech APC-65072-100UG
This item is not returnable.
View return policy
Description
The CD45.2 antibody from Proteintech is a mouse monoclonal antibody generated with b10.s mouse thymocytes and splenocytes of mouse CD45.2. This antibody recognizes mouse antigen. The CD45.2 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
CD45.2 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
Primary | |
Mouse |
Flow Cytometry | |
104 | |
APC | |
100 μg | |
19264 | |
IgG2a |