missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Mouse, PE, Clone: UCHT2, enQuireBio™
Mouse Monoclonal Antibody
119.00 € - 240.00 €
Specifications
Antigen | CD5 |
---|---|
Clone | UCHT2 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961548
|
EnquireBio
QAB15-PE-25Tests |
25 Tests |
119.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15951548
|
EnquireBio
QAB15-PE-100Tests |
100 Tests |
240.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD5 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
921 | |
IgG1 κ | |
Protein A/G Purified |
UCHT2 | |
Flow Cytometry | |
PE | |
RUO | |
NP_055022 | |
CD5 | |
Primary | |
Store at 2-4°C. |