missing translation for 'onlineSavingsMsg'
Learn More

CD56/NCAM-1 Ab-4 Mouse Monoclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD56/NCAM-1 Ab-4, Mouse Monoclonal Antibody.

Brand:  Epredia MS-1149-P0

 View more versions of this product

Product Code. 12679096

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135 kDa isoform is the basic molecule which is glycosylated or sialylated to produce the mature species. NCAM (CD56) is reported to express on most neuroectodermal derived cell lines, tissues, and neoplasms such as retinoblastoma, medullblastoma, astrocytoma, and neuroblastoma. It is also expressed on some mesodermally derived tumors such as rhabdomyosarcoma and also on natural killer cells.

Host Species: Mouse

Clone: 56C04; same as 123A8

Isotype: IgG1/κ

Species Reactivity: Human. Others not known

Epitope: Not determined

Immunogen: Membrane preparation of a small cell lung carcinoma

Molecular Weight: 180, 145 and 125 kDa

Positive Control: Neuroblastoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD56 Ab-4
Monoclonal
200μg/mL
Mouse
100 μL
Cell Adhesion
Human
IgG2a κ
Immunohistochemistry (Paraffin)
56C04; same as 123A8
Unconjugated
Membrane preparation of a small cell lung carcinoma
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.