missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68 Mouse anti-Human, Unconjugated, Clone: Y1/82A, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65187-1-IG-100UG
This item is not returnable.
View return policy
Description
CD68 is a type I transmembrane glycoprotein that is highly expressed by human monocytes and tissue macrophages. It belongs to the lysosomal/endosomal-associated membrane glycoprotein (LAMP) family and primarily localizes to lysosomes and endosomes with a smaller fraction circulating to the cell surface. CD68 is also a member of the scavenger receptor family. It may play a role in phagocytic activities of tissue macrophages.
Specifications
CD68 | |
Monoclonal | |
1 mg/mL | |
Mouse | |
Primary | |
Human | |
IgG2a |
Flow Cytometry | |
Y1/82A | |
Unconjugated | |
100 μg | |
968 | |
Liquid |