missing translation for 'onlineSavingsMsg'
Learn More

CD71 Rat, anti-Mouse, Clone: RI7217, Proteintech

Rat Monoclonal Antibody

Brand:  Proteintech 65289-1-IG-500UG

 View more versions of this product

Product Code. 18069415

  • 121.00 € / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The CD71 antibody from Proteintech is a rat monoclonal antibody generated with dmso induced friend erythroleukemia 745.6 of mouse CD71. This antibody recognizes mouse antigen. The CD71 antibody has been validated for the following applications: FC analysis.

Specifications

Specifications

CD71
RI7217
2610028K12Rik, AI195355, AI426448, AU015758, CD71, E430033M20Rik, Mtvr 1, Mtvr1, p90, TFR, TFR1, Tfrc, transferrin receptor, Trfr
500 μg
22042
Store at 2°C to 8°C. Stable for one year after shipment.
Liquid
Flow Cytometry
0.5 mg/ml
Rat
Primary
Mouse
150 kDa
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.