missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8 Mouse, PerCP, Clone: SK1, enQuireBio™
Mouse Monoclonal Antibody
252.00 € - 2660.00 €
Specifications
Antigen | CD8 |
---|---|
Clone | SK1 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941598
|
EnquireBio
QAB18-PCP-25Tests |
25 Tests |
252.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15931598
|
EnquireBio
QAB18-PCP-100Tests |
100 Tests |
605.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15951598
|
EnquireBio
QAB18-PCP-500Tests |
500 Tests |
2660.00 €
500 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
CD8 | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
925 | |
IgG1 κ | |
Protein A/G Purified |
SK1 | |
Flow Cytometry | |
PerCP | |
RUO | |
NP_741969 | |
CD8A | |
Primary | |
Store at 2-4°C. |