missing translation for 'onlineSavingsMsg'
Learn More

CD8a Mouse, APC-Cy7, Clone: RPA-T8, enQuireBio™

Mouse Monoclonal Antibody

143.00 € - 216.00 €

Specifications

Antigen CD8a
Clone RPA-T8
Concentration 5μL/Test
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15971608
View Documents Promotion Details
EnquireBio
QAB19-APC7-25Tests
25 Tests
143.00 €
25 tests
Please to purchase this item. Need a web account? Register with us today!
15961608
View Documents Promotion Details
EnquireBio
QAB19-APC7-100Tests
100 Tests
216.00 €
100 tests
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD8a
5μL/Test
Monoclonal
Mouse
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
925
IgG1 κ
Protein A/G Purified
RPA-T8
Flow Cytometry
APC-Cyanine7
RUO
NP_741969
CD8A
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications