missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD99 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547551
This item is not returnable.
View return policy
Description
In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) PILR- alpha and rmPILR- beta is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CD99 Polyclonal antibody specifically detects CD99 in Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
CD99 | |
Polyclonal | |
Unconjugated | |
CD99 | |
1110061M03Rik; 12E7; 2410026K10Rik; antigen identified by monoclonal 12E7, Y homolog; antigen identified by monoclonal antibodies 12E7, F21 and O13; Cd99; CD99 antigen; CD99 molecule; cell surface antigen 12E7; cell surface antigen HBA-71; cell surface antigen O13; D4; E2 antigen; HBA71; MIC2; MIC2 (monoclonal antibody 12E7); MIC2X; MIC2Y; MSK5X; paired immunoglobin-like type 2 receptor-ligand; pilr-1; Pilrl; PILR-L; Protein MIC2; surface antigen MIC2; T-cell surface glycoprotein E2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
673094 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8VCN6 | |
CD99 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD99 Gly25-Leu138 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction