missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC25B Mouse anti-Human, Unconjugated, Clone: 1H3A4, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | CDC25B |
---|---|
Clone | 1H3A4 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16809615
|
Proteintech
67145-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16899605
|
Proteintech
67145-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CDC25B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDC25B. This antibody recognizes human, mouse, rat antigen. The CDC25B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
CDC25B | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
994 | |
Primary |
1H3A4 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2b |