Learn More
CDK6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14052-1-AP-20UL
 View more versions of this product
Product Code. 16812524
  • 145.00 € / 20µL
Explore more special offers
Description
Description
IARS2 belongs to the class-I aminoacyl-tRNA synthetase family. Members of class I have two highly conserved sequence motifs. They aminoacylate at the 2′-OH of an adenosine nucleotide, and are usually monomeric or dimeric (one or two subunits, respectively). Both classes of aminoacyl-tRNA synthetases are multidomain proteins. The catalytic domains of all the aaRSs of a given class are found to be homologous to one another, while class I and class II aaRSs are unrelated to one another. The class I aaRSs have the ubiquitous Rossmann fold and have the antiparallel beta-strands architecture while the class II aaRSs have a unique fold made up of antiparallel beta-strands.
Specifications
Specifications
CDK6
Polyclonal
Unconjugated
CDK6
CDK6, Cell division protein kinase 6, cyclin dependent kinase 6, PLSTIRE
Rabbit
Antigen Affinity Chromatography
RUO
1021, 114483, 12571
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q00534, Q64261
Cdk6
CDK6 Fusion Protein Ag5275
20 μL
Primary
Rat, Mouse, Human