missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CDK7/9tide Substrate

A peptide substrate based upon the C-terminal tail of RNA Polymerase II that is a reported substrate for Cyclin Dependent Kinases (CDK7, CDK8 and CDK9).

Brand:  Thermo Scientific™ PV5090

Product Code. 10754511

  • 890.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

A peptide substrate based upon the C-terminal tail of RNA Polymerase II that is a reported substrate for Cyclin Dependent Kinases (CDK7, CDK8 and CDK9). For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

YSPTSPSYSPTSPSYSPTSPSKKKK
1mL
Fluorescence
1 mL
Peptide-Based Substrate
Serine/Threonine Kinases
Cdk7/9tide Substrate, 1mL, 2mM Concentration, 2689.9Da Molecular Weight, Peptide Substrate Based upon the C-terminal Tail of RNA Polymerase II that is a Reported Substrate For Cyclin Dependent Kinases (CDK7, CDK8 and CDK9)
CDK7/9tide Substrate
–20°C
CDK7, CDK9
Biochemical ADP accumulation
2 mM CDK7/9tide peptide substrate in 50 mM HEPES (pH 7.5), 0.01% Brij-35, 10 mM MgCl2 and 1 mM EGTA.

Store at -20°C.
Pharma and biopharma, protein assays and analysis, target and lead identification and validation, Adapta™ Universal Kinase Assay (PV5099), drug discovery and development, kinase biology, biochemical kinase assays, Adapta™ universal kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity
peptide based upon the C-terminal tail of RNA Polymerase II
Kinase Substrate
1022, 3376
2689.9 Da
Dry Ice
CDK9/Cyclin T1, CDK7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.